missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Vinyl bromide, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
$160.84 - $160.84
Specifications
Chemical Name or Material | Vinyl bromide |
---|---|
Name Note | 1M solution in THF |
CAS | 593-60-2,109-99-9 |
Density | 0.9270g/mL |
Flash Point | −17°C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC428771000
|
Thermo Scientific Chemicals
428771000 |
100 mL | AcroSeal™ Glass Bottle |
Each for $160.84
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Vinyl bromide | |
593-60-2,109-99-9 | |
−17°C | |
C2H3Br | |
MFCD00000183 | |
vinyl bromide, bromoethylene, ethene, bromo, monobromoethylene, vinylbromid, bromure de vinyle, vinylbromide, polyvinyl bromide, ethylene, bromo, polyvinylbromide | |
BrC=C | |
106.95 | |
CHEBI:51311 |
1M solution in THF | |
0.9270g/mL | |
AcroSeal™ Glass Bottle | |
H2C=CHBr | |
0.927 | |
INLLPKCGLOXCIV-UHFFFAOYSA-N | |
bromoethene | |
11641 | |
106.95 |
RUO – Research Use Only