missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vinyl iodide, tech. 85%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1265603

 View more versions of this product

Catalog No. AAL1265603


Explore available promotions

Description

Description

Vinyl iodide is an important intermediate used in organic synthesis, chemical research and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vinyl iodide is an important intermediate used in organic synthesis, chemical research and pharmaceuticals.

Solubility
Not miscible or difficult to mix in water.

Notes
It is sensitive to light. Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
Specifications

Specifications

Vinyl iodide
2.08
C2H3I
MFCD00039404
1731578
vinyl iodide, ethene, iodo, iodoethylene, vinyl iodide, tech., 1-iodoethylene, iodoethyl, iodo-ethene, 1-iodoethyl, 1-iodoethylene #, vinyl iodide, tech
GHXZPUGJZVBLGC-UHFFFAOYSA-N
iodoethene
68976
153.95
Technical
593-66-8
55°C to 56°C
1.53
1 g
Light sensitive
Not miscible or difficult to mix in water.
IC=C
153.95
CHEBI:51315
85%

Chemical Identifiers

593-66-8
153.95
GHXZPUGJZVBLGC-UHFFFAOYSA-N
68976
iodoethene
C2H3I
MFCD00039404
vinyl iodide, ethene, iodo, iodoethylene, vinyl iodide, tech., 1-iodoethylene, iodoethyl, iodo-ethene, 1-iodoethyl, 1-iodoethylene #, vinyl iodide, tech
CHEBI:51315
IC=C
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H302
May cause cancer.
Harmful if swallowed.

P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c

H302-H350

EINECSNumber : 209-801-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only