missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vinylbenzyl trimethylammonium chloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 422560250

 View more versions of this product

Catalog No. AC422560250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Phase transfer catalyst
Specifications

Specifications

Vinylbenzyl trimethylammonium chloride
Authentic
Glass bottle
CH2CHC6H4CH2N(CH3)3Cl
25g
211.73
26616-35-3
96% min. (Argentometry)
C12H18ClN
MFCD00080690
211.73
97%

Chemical Identifiers

26616-35-3
C12H18ClN
211.73
MFCD00080690
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS Signal Word: Warning

EINECSNumber : 247-850-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only