Learn More
Vinyltrimethoxysilane, 98%
CAS: 2768-02-7 | C5H12O3Si | 148.24 g/mol
$99.60 - $99.60
Chemical Identifiers
| CAS | 2-7-2768 |
|---|---|
| Molecular Formula | C5H12O3Si |
| Molecular Weight (g/mol) | 148.24 |
| MDL Number | MFCD00008605 |
| InChI Key | NKSJNEHGWDZZQF-UHFFFAOYSA-N |
| Synonym | vinyltrimethoxysilane, trimethoxyvinylsilane, silane, ethenyltrimethoxy, trimethoxy vinyl silane, trimethoxysilyl ethene, vinyl trimethoxy silane, silane, trimethoxyvinyl, trimethoxysilyl ethylene, vtmo, vts-m |
| PubChem CID | 76004 |
| IUPAC Name | ethenyl(trimethoxy)silane |
| SMILES | CO[Si](C=C)(OC)OC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC216521000
|
Thermo Scientific Chemicals
216521000 |
100 g | Glass bottle |
Each for $99.60
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2-7-2768 | |
| 148.24 | |
| NKSJNEHGWDZZQF-UHFFFAOYSA-N | |
| 76004 | |
| CO[Si](C=C)(OC)OC |
| C5H12O3Si | |
| MFCD00008605 | |
| vinyltrimethoxysilane, trimethoxyvinylsilane, silane, ethenyltrimethoxy, trimethoxy vinyl silane, trimethoxysilyl ethene, vinyl trimethoxy silane, silane, trimethoxyvinyl, trimethoxysilyl ethylene, vtmo, vts-m | |
| ethenyl(trimethoxy)silane |
Specifications
| 2-7-2768 | |
| 123.0°C | |
| 97.5% min. (GC) | |
| C5H12O3Si | |
| H2C=CHSi(OCH3)3 | |
| 100 g | |
| vinyltrimethoxysilane, trimethoxyvinylsilane, silane, ethenyltrimethoxy, trimethoxy vinyl silane, trimethoxysilyl ethene, vinyl trimethoxy silane, silane, trimethoxyvinyl, trimethoxysilyl ethylene, vtmo, vts-m | |
| NKSJNEHGWDZZQF-UHFFFAOYSA-N | |
| ethenyl(trimethoxy)silane | |
| 76004 | |
| 148.24 | |
| Vinyltrimethoxysilane |
| 0.9680g/mL | |
| 23°C | |
| Glass bottle | |
| 1.3915 to 1.3935 | |
| MFCD00008605 | |
| 0.968 | |
| Solubility in water: slightly soluble. Other solubilities: soluble in most common organic solvents | |
| CO[Si](C=C)(OC)OC | |
| 148.24 | |
| 0.6 mm2/s (25°C) | |
| 98% |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest
GHS Signal Word: Warning
EINECSNumber : 220-449-8
RUO – Research Use Only