missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Scigen VIP-Fixative Leak-Proof Specimen Containers Neutral Buffered 10% Formalin Filled

Safely transport, store and retrieve pathology specimens using Scigen’s Forma-Vial™ system. Containers from 7 mL to 7.5 Liters, filled with 10% neutral buffered formalin, OSHA compliant.

Supplier:  Scigen SGN1480

 View more versions of this product

Catalog No. 23-730-579


Explore available promotions

Description

Description

  • Forma-Vial™ specimen containers are uniquely designed to be leak proof and protect against biohazardous or hazardous material release to the environment
  • Highly visible OSHA compliant labeling
  • Sizes range from 7mL dermatology containers to 7.5 Liters operating room organ buckets
  • Container openings range from 13mm to 300mm
  • Filled with 10% neutral buffered formalin to 50% of container’s capacity
  • Formalin waste designed to be treated in bulk using Neutralex or treated individually using Neutralexr tablets (patent pending).
  • Nonsterile containers marked with a lot code and expiration date, 4 year shelf life
  • Manufactured using ISO 13485 and FDA quality systems

Specifications

Specifications

Specimen Container Vial
No
Non-sterile
480 mL, pre-filled 50%
Polypropylene
Lined Cap
Specimen Container Pre-filled With Neutral Buffered 10% Formalin
Histology/cytology
98 mm
OSHA Compliant
9/Pk.
Yes
Polypropylene
16 oz., pre-filled 50%
White
89 mm
Translucent
84 mm
3.9 in.
50% Filled Neutral Buffered 10% Formalin
9/Cs.
Forma-Vial™ Specimen Containers
Videos
SDS
Documents

Documents

Promotions

Promotions