missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Viral Transport Medium Kit DFS Item

The Gibco Viral Transport Medium Kit produces a medium that is suitable for the transport of samples suspected to contain virus.

Supplier:  Gibco™ A48498

Catalog No. A48498

Explore available promotions

Description

Description

The Gibco Viral Transport Medium Kit produces a medium that is suitable for the transport of samples suspected to contain virus. The kit includes Hanks Balanced Salt Solution (1X, Cat. No. 14025076), heat-inactivated FBS (Cat. No. 16140089), gentamicin sulfate (50 mg/mL, Cat. No. 15750078) and amphotericin B (250 μg/mL, Cat. No. 15290026). Once these components are combined, the resulting Viral Transport Medium helps enable safe transport and storage of samples for viral testing. The kit contains sufficient components to produce 50 L of Viral Transport Medium.

Specifications

Specifications

• Hanks Balanced Salt Solution (50 x 1 L, Cat. No. 14025076)
• Heat-inactivated FBS (1000 mL, Cat. No. 16140089)
• Gentamicin sulfate (50 mg/mL, 10 x 10 mL, Cat. No. 15750078)
• Amphotericin B (250 μg/mL, 2 x 50 mL, Cat. No. 15290026)

1 X
Standard Serum Supplementation
No Glucose, No Glutamine, No HEPES, No Phenol Red, No Sodium Pyruvate
1 Kit
Animal Origin
Virus Transport Medium
Sterile-filtered
Kit

HBSS, Gentamicin: Room temperature
FBS, Amphotericin: Dry ice


Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.