VistaLab™ Technologies MLA™ Brand Digital Pipetters
Perfect for tests that require multiple or nonstandard volumes
 View more versions of this product
Catalog No. 21-369G
Explore available promotions
Description
Description
Three adjustable models cover volume range: 10 to 1000μL. To adjust volume, simply turn volume selector knob near top of pipetter. Four-digit reading is locked in by click stops, ensuring accuracy.
  • One-handed operation
  • D-Tipper™ feature allows you to pick up and dispose of tips without touching them
  • Volume range conveniently marked on all models
  • Factory calibrated for accurate delivery
  • Easy in-lab recalibration
  • Made of lightweight aluminum and stainless steel to assure durability and long life
Order Info
See MLA Brand tips Mfr. Nos. 2025, 2026, 2027, 2028, 4025, 4026, 9025, 9026, 9029
Specifications
Specifications
±1%
Not Autoclavable
0.2 mL Tip
1
Non-sterile
50 to 200 μL
0.2 mL Tip
1 Ea.
9 in.
1.5 in.
0.2 mL Tip
Red
Aluminum, Stainless Steel
MLA™ Brand
Digital Pipetter
Multiple or Non-Standard Volumes
Nozzle Insert, Calibration Key
Limited Lifetime
2 in.