Newport™ Vistek™ Vibration Isolation Platforms DFS Item
Significantly improve the performance of microscopes, lab balances, and other sensitive instruments
Supplier:  Newport™ LIP275122201
Catalog No. 10-999-633
Explore available promotions
Description
Description
Cut off the transmission of vibrations from the tabletop or countertop. Effectively isolate horizontal and vertical vibration components in all six degrees of freedom.
  • Passive, mechanical and maintenance-free No air hoses or supporting hardware required