missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Vitamin A Acetate, USP, Spectrum™ Chemical
V1257, 127-47-9, C22H32O2
$721.37 - $16938.20
Chemical Identifiers
CAS | 127-47-9 |
---|---|
Molecular Formula | C22H32O2 |
Molecular Weight (g/mol) | 328.50 |
InChI Key | QGNJRVVDBSJHIZ-QHLGVNSISA-N |
IUPAC Name | (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-yl acetate |
SMILES | CC(=O)OC\C=C(/C)\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18607708
|
Spectrum Chemical Mfg Cor
V1257100GM |
100 g | Amber Glass Bottle |
Each for $721.37
|
|
||||
18607709
|
Spectrum Chemical Mfg Cor
V1257500GM |
500 g | Amber Glass Bottle |
Each for $2,668.17
|
|
||||
18607710
|
Spectrum Chemical Mfg Cor
V125725KGBL |
25 kg | Fiber Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Vitamin A Acetate, USP is also known as retinyl acetate or retinol acetate and is often used to fortify foods with vitamin A. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
127-47-9 | |
328.50 | |
(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-yl acetate |
C22H32O2 | |
QGNJRVVDBSJHIZ-QHLGVNSISA-N | |
CC(=O)OC\C=C(/C)\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C |
Specifications
127-47-9 | |
Amber Glass Bottle | |
100 g | |
CC(=O)OC\C=C(/C)\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C | |
328.50 |
1 | |
C22H32O2 | |
QGNJRVVDBSJHIZ-QHLGVNSISA-N | |
(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-yl acetate | |
USP |