missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Vitamin B12 95.0+%, TCI America™
$61.22 - $203.43
Chemical Identifiers
CAS | 68-19-9 |
---|---|
Molecular Formula | C63H88CoN14O14P |
Molecular Weight (g/mol) | 1355.39 |
MDL Number | MFCD00151092 |
InChI Key | YUWVGNQGDAPKCX-BVWPOUIRNA-L |
Synonym | vitamin b12 |
PubChem CID | 129893524 |
IUPAC Name | (10S,12R,13R,24S,35R,41R)-30,35,40-tris(2-carbamoylethyl)-24,36,41-tris(carbamoylmethyl)-1-cyano-46-hydroxy-12-(hydroxymethyl)-5,6,17,23,28,31,31,36,38,41,42-undecamethyl-15,20-dioxo-11,14,16-trioxa-2λâµ,9,19,26λâµ,43,44λâµ,45λâµ-heptaaza-15λâµ-phospha-1-cobaltadodecacyclo[27.14.1.1¹,³â´.1²,â¹.1¹â°,¹³.0¹,²â¶.0³,â¸.0²³,²â·.0²âµ,â´Â².0³²,â´â´.0³â¹,â´Â³.0³â·,â´âµ]heptatetraconta-2(47),3,5,7,26,28,32(44),33,37(45),38-decaene-1,2,26,44,45-pentakis(ylium)-15-olate |
SMILES | [C@@H]1(C2=CC3=[N+]4C(C(C3(C)C)CCC(N)=O)=C(C3=[N+]5C6C7(N8C(C([C@@]7(C)CC(N)=O)CCC(N)=O)=C(C(=[N+]2[Co+]458([N+]2=CN(C4=CC(=C(C)C=C24)C)[C@@]2([H])O[C@@H]([C@]([H])(OP([O-])(=O)OC(C)CNC(=O)CCC3(C)[C@@H]6CC(N)=O)C2O)CO)C#N)C1(C)CC(N)=O)C)C)C)CCC(N)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
C0449100MG
|
TCI America
C0449100MG |
100 mg |
Each for $61.22
|
|
|||||
C04491G
|
TCI America
C04491G |
1 g |
Each for $203.43
|
|
|||||
Chemical Identifiers
68-19-9 | |
1355.39 | |
YUWVGNQGDAPKCX-BVWPOUIRNA-L | |
129893524 | |
[C@@H]1(C2=CC3=[N+]4C(C(C3(C)C)CCC(N)=O)=C(C3=[N+]5C6C7(N8C(C([C@@]7(C)CC(N)=O)CCC(N)=O)=C(C(=[N+]2[Co+]458([N+]2=CN(C4=CC(=C(C)C=C24)C)[C@@]2([H])O[C@@H]([C@]([H])(OP([O-])(=O)OC(C)CNC(=O)CCC3(C)[C@@H]6CC(N)=O)C2O)CO)C#N)C1(C)CC(N)=O)C)C)C)CCC(N)=O |
C63H88CoN14O14P | |
MFCD00151092 | |
vitamin b12 | |
(10S,12R,13R,24S,35R,41R)-30,35,40-tris(2-carbamoylethyl)-24,36,41-tris(carbamoylmethyl)-1-cyano-46-hydroxy-12-(hydroxymethyl)-5,6,17,23,28,31,31,36,38,41,42-undecamethyl-15,20-dioxo-11,14,16-trioxa-2λâµ,9,19,26λâµ,43,44λâµ,45λâµ-heptaaza-15λâµ-phospha-1-cobaltadodecacyclo[27.14.1.1¹,³â´.1²,â¹.1¹â°,¹³.0¹,²â¶.0³,â¸.0²³,²â·.0²âµ,â´Â².0³²,â´â´.0³â¹,â´Â³.0³â·,â´âµ]heptatetraconta-2(47),3,5,7,26,28,32(44),33,37(45),38-decaene-1,2,26,44,45-pentakis(ylium)-15-olate |
Specifications
68-19-9 | |
C63H88CoN14O14P | |
100 mg | |
YUWVGNQGDAPKCX-BVWPOUIRNA-L | |
(10S,12R,13R,24S,35R,41R)-30,35,40-tris(2-carbamoylethyl)-24,36,41-tris(carbamoylmethyl)-1-cyano-46-hydroxy-12-(hydroxymethyl)-5,6,17,23,28,31,31,36,38,41,42-undecamethyl-15,20-dioxo-11,14,16-trioxa-2λâµ,9,19,26λâµ,43,44λâµ,45λâµ-heptaaza-15λâµ-phospha-1-cobaltadodecacyclo[27.14.1.1¹,³â´.1²,â¹.1¹â°,¹³.0¹,²â¶.0³,â¸.0²³,²â·.0²âµ,â´Â².0³²,â´â´.0³â¹,â´Â³.0³â·,â´âµ]heptatetraconta-2(47),3,5,7,26,28,32(44),33,37(45),38-decaene-1,2,26,44,45-pentakis(ylium)-15-olate | |
129893524 | |
≥95.0% (E) | |
Vitamin B12 |
Purple-Red | |
MFCD00151092 | |
vitamin b12 | |
[C@@H]1(C2=CC3=[N+]4C(C(C3(C)C)CCC(N)=O)=C(C3=[N+]5C6C7(N8C(C([C@@]7(C)CC(N)=O)CCC(N)=O)=C(C(=[N+]2[Co+]458([N+]2=CN(C4=CC(=C(C)C=C24)C)[C@@]2([H])O[C@@H]([C@]([H])(OP([O-])(=O)OC(C)CNC(=O)CCC3(C)[C@@H]6CC(N)=O)C2O)CO)C#N)C1(C)CC(N)=O)C)C)C)CCC(N)=O | |
1355.39 | |
1355.39 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : GG3750000
TSCA : Yes
Recommended Storage : Refrigerator