missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Vitamin D2, 97+%

Vitamin D2, or Ergocalciferol, CAS # 50-14-6, a fat-soluble hormone that is important for calcium and phosphorus absorption and metabolism.

Supplier:  Thermo Scientific Chemicals 422630050

 View more versions of this product

Catalog No. AC422630050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Vitamin D2 (ergocalciferol) is a form of vitamin D, a fat-soluble hormone found in food sources
  • Vitamin D is important for calcium and phosphorus homeostasis, and can play a role in many biological processes including cell growth
  • Vitamin D2 has a lower binding affinity than Vitamin D3 for vitamin D binding protein

Applications

  • Research studies show that Vitamin D2 treatment in rats inhibits bladder tumor promotion
  • Studies show that the treatment of lung carcinoma cells with vitamin D2 in vitro resulted in increased ACE2 protein levels
  • Vitamin D2 has been seen to interfere with prion protein oligomerization in vitro
Specifications

Specifications

Vitamin D2
50-14-6
115.0°C to 119.0°C
Authentic
Glass bottle
MFCD00166988,MFCD11656674
15, 10215
+ 82.00
Solubility in water: insoluble. Other solubilities: soluble in common organic solvents
CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C
396.66
396.65
Powder
99%
460 to 490 (265nm, EtOH 96%)
Beige to White
97.0 to 103% (HPLC)
C28H44O
5 g
+82.00 (25.00°C c=3,in acetone)
ergocalciferol, vitamin d2, calciferol, viosterol, oleovitamin d2, ercalciol, ergorone, detalup, condocaps, crystallina
MECHNRXZTMCUDQ-VLOQVYPSSA-N
(1S)-3-[2-[(1S,7aR)-1-[(2R,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
134129658
97+%

Chemical Identifiers

50-14-6
396.66
MECHNRXZTMCUDQ-VLOQVYPSSA-N
134129658
CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C
C28H44O
MFCD00166988,MFCD11656674
ergocalciferol, vitamin d2, calciferol, viosterol, oleovitamin d2, ercalciol, ergorone, detalup, condocaps, crystallina
(1S)-3-[2-[(1S,7aR)-1-[(2R,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air

GHS Signal Word: Danger

EINECSNumber : 200-014-9

RTECSNumber : KE1050000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only