missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Vitamin K1, MP Biomedicals™
$359.00 - $359.00
Chemical Identifiers
CAS | 84-80-0 |
---|---|
Molecular Formula | C31H46O2 |
Molecular Weight (g/mol) | 450.707 |
InChI Key | MBWXNTAXLNYFJB-NKFFZRIASA-N |
Synonym | vitamin k1, phytonadione, phylloquinone, phytylmenadione, phyllochinon, phytomenadione, alpha-phylloquinone, 3-phytylmenadione, konakion, 2-methyl-3-phytyl-1,4-naphthoquinone |
PubChem CID | 5284607 |
ChEBI | CHEBI:18067 |
IUPAC Name | 2-methyl-3-[(E,7R,11R)-3,7,11,15-tetramethylhexadec-2-enyl]naphthalene-1,4-dione |
SMILES | CC1=C(C(=O)C2=CC=CC=C2C1=O)CC=C(C)CCCC(C)CCCC(C)CCCC(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN10328301
|
MP Biomedicals Inc
0210328301 |
1 g |
Each for $359.00
|
|
|||||
Chemical Identifiers
84-80-0 | |
450.707 | |
vitamin k1, phytonadione, phylloquinone, phytylmenadione, phyllochinon, phytomenadione, alpha-phylloquinone, 3-phytylmenadione, konakion, 2-methyl-3-phytyl-1,4-naphthoquinone | |
CHEBI:18067 | |
CC1=C(C(=O)C2=CC=CC=C2C1=O)CC=C(C)CCCC(C)CCCC(C)CCCC(C)C |
C31H46O2 | |
MBWXNTAXLNYFJB-NKFFZRIASA-N | |
5284607 | |
2-methyl-3-[(E,7R,11R)-3,7,11,15-tetramethylhexadec-2-enyl]naphthalene-1,4-dione |
Specifications
84-80-0 | |
0.984 | |
1 g | |
MBWXNTAXLNYFJB-NKFFZRIASA-N | |
2-methyl-3-[(E,7R,11R)-3,7,11,15-tetramethylhexadec-2-enyl]naphthalene-1,4-dione | |
5284607 | |
450.68 | |
Vitamin K1 |
Yellow | |
C31H46O2 | |
vitamin k1, phytonadione, phylloquinone, phytylmenadione, phyllochinon, phytomenadione, alpha-phylloquinone, 3-phytylmenadione, konakion, 2-methyl-3-phytyl-1,4-naphthoquinone | |
CC1=C(C(=O)C2=CC=CC=C2C1=O)CC=C(C)CCCC(C)CCCC(C)CCCC(C)C | |
450.707 | |
CHEBI:18067 | |
Viscous Liquid |