missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Vitronectin (VTN-N) Recombinant Human Protein, Truncated
Vitronectin (VTN-N) is a recombinant human protein that provides a defined surface for feeder-free culture of human pluripotent stem cells (PSCs). When used with Essential 8™ Medium (A1517001), VTN-N has been proven to maintain pluripotency and normal growth characteristics in multiple PSC lines.
$126.00 - $1208.00
Specifications
Concentration | 500 μg⁄ml |
---|---|
For Use With (Application) | Stem Cell Culture |
Species | Human |
Recombinant | Recombinant |
Content And Storage | Store at -80°C and protect from light. |
Description
- When used with Essential 8™ Medium (A1517001), vitronectin (VTN-N) has been proven to maintain pluripotency and normal growth characteristics in multiple PSC lines.
- Supports PSC growth for >50 passages without any signs of karyotypic abnormalities and to maintain the ability of PSCs to differentiate into all three germ line lineages.
- Supports human pluripotent stem cell attachment and survival better than wild-type vitronectin when used in conjunction with Essential 8™ Medium.
- Optimized—designed in the laboratory of James Thomson for use with Essential 8™ Medium
- Defined—for reduced variability in your PSC cultures
- Support human pluripotent stem cell attachment and survival better than wild type vitronectin.
- Optimal environment for feeder-free pluripotent stem cell culture.
- Defined, recombinant human protein that reduces variability in your PSC cultures compared to human-plasma derived vitronectin and standard basement membrane extracts (BMEs).
Specifications
500 μg⁄ml | |
Human | |
Store at -80°C and protect from light. | |
Liquid | |
Stem Cell Research |
Stem Cell Culture | |
Recombinant | |
Recombinant | |
Vitronectin |
For Research Use Only. Not for use in diagnostic procedures.