Learn More
Cytiva Vivaspin™ 20 Sample Concentrators
Use for one-step, non-denaturing sample concentration in a single tube for minimal sample handling and reduced sample loss. Cytiva Life Vivaspin™ 20 Sample Concentrators cater to scientists working with sample volumes ranging from 100 μl to 20 ml, with a range of MWCO values from Mr 3 000 to 100 000.
$263.51 - $267.83
Specifications
Material (Membrane) | PES |
---|---|
Filtration Area | 6 cm2 |
Volume (Metric) | 5 to 20 mL |
Length (Metric) | 116 mm |
Width (Metric) | 30 mm |
Catalog Number | Mfr. No. | MWCO | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | MWCO | Price | Quantity | |||||
45-001-580
|
Cytiva
28932361 |
30 kDa |
Pack of 12 Each for $263.51
|
|
|||||
45-001-577
|
Cytiva
28932358 |
3 kDa |
Pack of 12 Each for $267.83
|
|
|||||
45-001-578
|
Cytiva
28932359 |
5 kDa |
Pack of 12 Each for $264.51
|
|
|||||
45-001-581
|
Cytiva
28932362 |
50 kDa |
Pack of 12 Each for $264.51
|
|
|||||
45-001-582
|
Cytiva
28932363 |
100 kDa |
Pack of 12 Each for $264.51
|
|
|||||
45-001-579
|
Cytiva
28932360 |
10 kDa |
Pack of 12 Each for $263.51
|
|
|||||
Description
Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%. Patented dead-stop technology ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery.
- Accept sample volumes from 100μLto 20mL, with a range of molecular weight cutoff values from Mr3000 to 100,000
- Vertical polyethersulfone semi-permeable membrane separating upper and lower chamber resists blockage and tolerates high flow rate
- Centrifugation forces solvent through membrane, leaving concentrated sample in the upper chamber
- Compatible pH range: 1 to 9
- Active membrane area: 6.0cm2
For maximum recovery, select a MWCO at least 50% smaller than the molecular size of the species of interest.
Specifications
PES | |
5 to 20 mL | |
30 mm |
6 cm2 | |
116 mm |