missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sartorius Vivaspin™ Turbo 4 Ultrafiltration Unit
Handles up to 4mL sample volume in 4 swing-out and fixed angle rotors accepting 15mL centrifuge tubes. Sartorius Vivaspin Turbo 4 Ultrafiltration Unit, features twin vertical membranes and sleek internal design for fastest sample concentration with highest recoveries.
$250.54 - $881.53
Specifications
Description | Turbo 4 Ultrafiltration Unit |
---|---|
Material (Membrane) | PES |
For Use With (Application) | Concentration, Desalting, buffer exchange |
Max. Force | Swing-out rotor : 4000 x gAngle rotor : 7500 x g |
Sample Type | Sample type: Nanoparticles; Peptide; Protein |
Catalog Number | Mfr. No. | MWCO | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | MWCO | Quantity | Price | Quantity | ||||
14-558-703
|
Sartorius
VS04T91 |
3 kDa | 25/Pk. |
N/A
|
|
||||
14-558-704
|
Sartorius
VS04T92 |
3 kDa | 100/Pk. |
N/A
|
|
||||
14-558-705
|
Sartorius
VS04T11 |
5 kDa | 25/Pk. |
N/A
|
|
||||
14-558-706
|
Sartorius
VS04T12 |
5 kDa | 100/Pk. |
N/A
|
|
||||
14-558-707
|
Sartorius
VS04T01 |
10 kDa | 25/Pk. |
N/A
|
|
||||
14-558-708
|
Sartorius
VS04T02 |
10 kDa | 100/Pk. |
N/A
|
|
||||
14-558-709
|
Sartorius
VS04T21 |
30 kDa | 25/Pk. |
N/A
|
|
||||
14-558-710
|
Sartorius
VS04T22 |
30 kDa | 100/Pk. |
N/A
|
|
||||
14-558-711
|
Sartorius
VS04T31 |
50 kDa | 25/Pk. |
N/A
|
|
||||
14-558-712
|
Sartorius
VS04T32 |
50 kDa | 100/Pk. |
N/A
|
|
||||
14-558-713
|
Sartorius
VS04T41 |
100 kDa | 25/Pk. |
N/A
|
|
||||
14-558-714
|
Sartorius
VS04T42 |
100 kDa | 100/Pk. |
N/A
|
|
||||
Description
Handles up to 4mL sample volume in 4 swing-out and fixed angle rotors accepting 15mL centrifuge tubes. Sartorius Vivaspin Turbo 4 Ultrafiltration Unit, features twin vertical membranes and sleek internal design for fastest sample concentration with highest recoveries.
- High flux PES membrane
- Retentate recovery >95% x pH 1-14 suited polystyrene housing with high chemical compatibility
- Universal rotor compatibility
- Volume graduation for easy detection / monitoring of the concentrate volume
- UV joining technology allows for smooth joint transition between membrane and plastic housing, allowing collection of complete concentrated sample into unique pipette friendly dead stop pocket
Specifications
Turbo 4 Ultrafiltration Unit | |
Concentration, Desalting, buffer exchange | |
Sample type: Nanoparticles; Peptide; Protein | |
2 to 4 mL |
PES | |
Swing-out rotor : 4000 x gAngle rotor : 7500 x g | |
Centrifugal Unit | |
1225 mm |