Fisherbrand™ Vortex Mixers Tube Holders
For use with Fisherbrand vortex mixers
Supplier:  Fisherbrand™ 80863008
 View more versions of this product
Catalog No. 02-215-402
Explore available promotions
Description
Description
  • Mount atop mixer for single-tube, hands-free mixing
  • Easily attach without tools
Specifications
Specifications
0.5 mL
9 x 9 in. (229 x 229 mm)
229 mm
229 mm
24
Microtube Holder
9 in.
9 in.
Vortex mixer
Product Suggestions
Product Suggestions