Invitrogen™ Vybrant™ Apoptosis Assay Kits for apoptotic and necrotic cell staining DFS Item
Easily distinguish cells undergoing apoptosis with Vybrant Apoptosis Assay Kits, which offer dyes, including YO-PRO-1 and Hoechst 33342, for apoptotic and necrotic cell staining.
Supplier:  Invitrogen™ V23200
 View more versions of this product
Catalog No. 501121558
Explore available promotions
Description
Description
Apoptotic cells undergo several physiological processes, including changes in cell membrane permeability and chromatin condensation, which can be distinguished using different dyes in apoptotic and necrotic cell staining protocols. Vybrant Apoptosis Assay kits offer streptavidin biotin-annexin V, propidium iodide (PI), YO-PRO-1, and Hoechst 33342 dyes to differentiate live from apoptotic or necrotic cells, which can then be visualized using flow cytometry.
Vybrant Apoptosis Assay kits offer quick and convenient cell staining dye-based methodologies for differentiating apoptotic and necrotic versus healthy cells, which can then be visualized using flow cytometry.
The Vybrant Apoptosis Assay Kit #4 detects apoptosis via changes that occur in cell membrane permeability. This kit contains ready-to-use solutions of both YO-PRO-1 and propidium iodide (PI) nucleic acid stains. YO-PRO-1 stain selectively passes through the plasma membranes of apoptotic cells and labels them with moderate green fluorescence. Necrotic cells are stained fluorescent red with PI.
The Vybrant Apoptosis Assay Kit #5, or the Chromatin Condensation/Dead Cell Apoptosis Kit with Hoechst 33342 and PI for Flow Cytometry, provides a rapid and convenient assay for apoptosis based on fluorescence detection of the compacted state of the chromatin in apoptotic cells. This kit contains ready-to-use solutions of blue-fluorescent Hoechst 33342 dye, which stains the condensed chromatin of apoptotic cells more brightly than the chromatin of non-apoptotic cells, and red-fluorescent PI dye, which stains dead cells.
The Vybrant Apoptosis Assay Kit #6 provides a rapid and convenient assay for apoptosis, using the bright, blue-fluorescent Alexa Fluor 350 conjugate of streptavidin in conjunction with biotin-X annexin V to detect apoptotic cell populations either by flow cytometry or imaging. In addition, the kit includes a ready-to-use solution of the fluorescent red PI nucleic acid–binding dye. PI is impermeant to live cells and apoptotic cells, but stains dead cells with red fluorescence, binding tightly to their nucleic acid. The kit contains an ample amount of each reagent for about 50 flow cytometric assays.
Specifications
Specifications
Vybrant Apoptosis Assay Kit #6
Tube
Apoptosis Kit
Alexa Fluor™ 346/442: 346
PI: 535/617
Wet Ice
Fluorescence
Alexa Fluor™ 350 Streptavidin, propidium iodide, Biotin-X Annexin V
Store in refrigerator (2–8°C) and protect from light
50 assays
1 vial of biotin-x annexin V, 1 vial of Alexa Fluor™ 350 streptavidin, 1 vial of propidium iodide, and 1 bottle of 5x annexin binding buffer
UV, 488
Blue, Red
Alexa Fluor™
Flow Cytometer
Alexa Fluor™ Dyes, Biotin & Other Haptens, Other Label(s) or Dye(s)