Fisherbrand™ Walk-In Fume Hood Work Floor
Fisherbrand™ Walk-In Fume Hood Work Floor is designed to be used with Fisherbrand Walk-in Fume Hoods.
 View more versions of this product
Catalog No. 06000615
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • 1.25 in. thick modified black epoxy resin work floors are an option for all Walk-In Fume Hoods
  • Designed to retain spills.
Specifications
Specifications
Epoxy
31 in.
Fisherbrand Walk-In Fume Hoods
3.2 cm
182.88 cm
Black
78.74 cm
1.25 in.
72 in.