missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Warfarin Sodium (contains Isopropyl Alcohol) 98.0+%, TCI America™
$123.03 - $363.49
Chemical Identifiers
| CAS | 129-06-6 |
|---|---|
| Molecular Formula | C19H18NaO4 |
| Molecular Weight (g/mol) | 333.339 |
| MDL Number | MFCD00083223 |
| InChI Key | MBNHFFMQYKZCLK-UHFFFAOYSA-N |
| Synonym | 3-(alpha-Acetonylbenzyl)-4-hydroxycoumarin Sodiuim Salt, 4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin Sodium Salt |
| PubChem CID | 131664125 |
| IUPAC Name | 4-hydroxy-3-(3-oxo-1-phenylbutyl)chromen-2-one;molecular hydrogen;sodium |
| SMILES | [HH].CC(=O)CC(C1=CC=CC=C1)C2=C(C3=CC=CC=C3OC2=O)O.[Na] |
Chemical Identifiers
| 129-06-6 | |
| 333.339 | |
| MBNHFFMQYKZCLK-UHFFFAOYSA-N | |
| 131664125 | |
| [HH].CC(=O)CC(C1=CC=CC=C1)C2=C(C3=CC=CC=C3OC2=O)O.[Na] |
| C19H18NaO4 | |
| MFCD00083223 | |
| 3-(alpha-Acetonylbenzyl)-4-hydroxycoumarin Sodiuim Salt, 4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin Sodium Salt | |
| 4-hydroxy-3-(3-oxo-1-phenylbutyl)chromen-2-one;molecular hydrogen;sodium |
Specifications
| 129-06-6 | |
| C19H18NaO4 | |
| 5 g | |
| 3-(alpha-Acetonylbenzyl)-4-hydroxycoumarin Sodiuim Salt, 4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin Sodium Salt | |
| [HH].CC(=O)CC(C1=CC=CC=C1)C2=C(C3=CC=CC=C3OC2=O)O.[Na] | |
| 333.339 | |
| 330.31 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00083223 | |
| 1544 | |
| MBNHFFMQYKZCLK-UHFFFAOYSA-N | |
| 4-hydroxy-3-(3-oxo-1-phenylbutyl)chromen-2-one;molecular hydrogen;sodium | |
| 131664125 | |
| ≥98.0% (HPLC) | |
| Warfarin Sodium (contains Isopropyl Alcohol) |
Safety and Handling
RTECSNumber : GN4725000
TSCA : Yes