missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Conair™ Waring™ Laboratory Blenders: Two Speeds, Commercial
Commercial-grade units for demanding mixing and blending applications
Supplier: Conair™ WRG7011S
Includes: Base, container, 3-wire grounded cord and plug
Description
Commercial-grade units for demanding mixing and blending applications
- Cloverleaf design
- Choice of glass and/or stainless-steel containers
- Sturdy, diecast metal bases and stainless-steel blades
- All durably constructed for years of dependable service
- Epoxy-coated bases with chrome top pieces on all models, except for the aluminum and steel base (Hazardous-Location Model)
- Heat-resistant borosilicate glass (integral handle) or stainless-steel (attached handles)
- Two-piece vinyl and styrene lids on all models
Order Info
Electrical requirements for all models: 115/120V 50/60Hz
Certifications
UL listed (Select models only; see ordering chart)
Specifications
3.6 A | |
115/120 V | |
50/60 Hz | |
1000 mL | |
8.5 x 7.25 x 15 in. (21.59 x 18.41 x 38 cm) | |
38 cm | |
21.59 cm | |
18.41 cm | |
Stainless Steel | |
1 Ea. |
18,000, 22,000 rpm | |
115/120 V, 50/60 Hz | |
0.4 hp | |
34 oz. | |
15 in. | |
8.5 in. | |
7.25 in. | |
Epoxy-coated Metal | |
Base, container, 3-wire grounded cord and plug |