missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Conair™ Waring™ Laboratory Blenders: Two Speeds with Timer

Dual-speed action for demanding mixing and blending applications

Supplier:  Conair™ 7010S

 View more versions of this product

Catalog No. 14-509-7P


Explore available promotions

Includes: Base, container, 3-wire grounded cord and plug

Description

Description

  • Cloverleaf design
  • Choice of glass and/or stainless-steel containers
  • Sturdy, diecast metal bases and stainless-steel blades
  • All durably constructed for years of dependable service
  • Epoxy-coated bases with chrome top pieces on all models, except for the aluminum and steel base (Hazardous-Location Model)
  • Heat-resistant borosilicate glass (integral handle) or stainless-steel (attached handles)
  • Two-piece vinyl and styrene lids

Order Info

Electrical requirements for all models: 115/120V 50/60Hz

Certifications

UL listed (Select models only; see ordering chart)

Specifications

Specifications

3.6 A
115/120 V
50/60 Hz
1000 mL
9.75 x 8 x 15 in. (24.76 x 20.32 x 38 cm)
38 cm
24.76 cm
20.32 cm
Stainless Steel
1 Ea.
18,000, 22,000 rpm
115/120 V, 50/60 Hz
0.4 hp
34 oz.
15 in.
9.75 in.
8 in.
Epoxy-coated Metal
Base, container, 3-wire grounded cord and plug, timer
Videos
SDS
Documents

Documents

Promotions

Promotions