missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
ShockWatch™ WarmMark™ Time-Temperature Indicator
ShockWatch WarmMark Temperature Indicator is a single-use dacending temperature indicator, which alerts users of exposure to unacceptable temperature conditions.
$436.76 - $436.76
Specifications
Certifications/Compliance | ROHS2, REACH |
---|---|
Disposable | Yes |
For Use With (Application) | Shipping and storage |
Thermometer Fill | White to Red |
Product Type | Disposable Temperature Indicator |
Alert:
Contains no known hazardous or carcinogenic components
Catalog Number | Mfr. No. | Temperature Range (English) | Measurement Time | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Temperature Range (English) | Measurement Time | Price | Quantity | ||||
22-111-030
![]() |
ShockWatch™
WM8468 |
46°F | 8 hr. |
Pack of 100 Each for $436.76
|
|
||||
22111029
![]() |
ShockWatch™
WM846 |
46°F | 48 hr. |
Pack of 100 Each for $436.76
|
|
||||
22-111-031
![]() |
ShockWatch™
WM032 |
36°F |
Pack of 100 Each for $436.76
|
|
|||||
Description
- Field armable with pull tab
- Black stripes show product armed
- Brief, moderate, prolonged exposure times
Order Info
Store in dry environment below response temperature for optimal shelf life
Certifications
ROHS2, REACH
Warranty
2 years from date of sale
Specifications
ROHS2, REACH | |
Shipping and storage | |
Disposable Temperature Indicator |
Yes | |
White to Red | |
±1°C |