Cytiva Wash Valve
Supplier: Cytiva 29023897
Description
For use with the AKTA start automated protein purification system. The wash valve is used to divert the flow path to the waste when needed.Specifications
Divert the flow path to the waste when needed | |
AKTA start Automated Protein Purification System |