missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Water, 0.1% Trifluoroacetic Acid, BAKER ANALYZED™ LC-MS Reagent, J.T. Baker™
Function tested and optimized for LC/MS applications
Supplier: Avantor J.T.Baker 983603
Description
- For use in liquid chromatography and mass spectrometry; For laboratory, research, or manufacturing use
Specifications
Water | |
7732-18-5 | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
Narrow Mouth Amber Glass Bottle | |
1L = 1.00kg |
0.1% Trifluoroacetic Acid | |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
BAKER ANALYZED™ LC-MS Reagent | |
4 L |
Chemical Identifiers
7732-18-5 | |
18.015 | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
CHEBI:15377 | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |