missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mobile phase for HPLC, Water with trifluoroacetic acid (0.1%), HPLC Grade, Fisher Chemical
Supplier: Fisher Chemical HB5134
Description
- Ideal mobile phase blend developed for use with HPLC instruments
- Pre-mixed for ease-of-use
- TFA is a common mobile phase additive that can function as an ion pair reagent
Specifications
Water with 0.1% Trifluoroacetic Acid (v/v) | |
H2O | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
HPLC | |
4 L |
7732-18-5,76-05-1 | |
MFCD00011332 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
Inclusive between 0.095% | |
Glass Bottle | |
Liquid |
Chemical Identifiers
7732-18-5 | |
18.015 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
H2O | |
MFCD00011332 | |
dihydrogen oxide, dihydrogen monoxide | |
CHEBI:15377 | |
O |
Safety and Handling
CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:0
Instability:0