missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisher Science Education™ Water
Supplier: Fisher Science Education™ S25868
Alert:
This item is not for human consumption
Specifications
Water | |
100% w/v | |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02 | |
1.000 | |
Liquid |
7732-18-5 | |
H2O | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
Distilled | |
1 gal. |
Chemical Identifiers
7732-18-5 | |
18.015 | |
dihydrogen oxide, dihydrogen monoxide | |
CHEBI:15377 | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
Safety and Handling
ShelfLife : 5 years