missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Water (Biotech Grade), Fisher BioReagents
Supplier: Fisher BioReagents BP248520
Description
Biotech-grade water can be used for several biological test procedures.Specifications
Water | |
7732-18-5 | |
0°C | |
H2O | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
Biotech | |
17.5mmHg at 20°C | |
1.000g/cm³ | |
Liquid |
7 | |
Colorless | |
100°C | |
15, 10237 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02 | |
Poly CUBE | |
Filtered through a 0.2-micron filter. | |
20 L |
Chemical Identifiers
7732-18-5 | |
18.015 | |
dihydrogen oxide, dihydrogen monoxide | |
CHEBI:15377 | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
Safety and Handling
Low hazard for usual industrial or commercial handling.
Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult