missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water (Biotech Grade), Fisher BioReagents

Supplier:  Fisher BioReagents BP24854

 View more versions of this product

Catalog No. BP24854


Explore available promotions

Description

Description

Biotech-grade water can be used for several biological test procedures.
Specifications

Specifications

Water
7732-18-5
0°C
H2O
dihydrogen oxide, dihydrogen monoxide
O
18.015
CHEBI:15377
Biotech
17.5mmHg at 20°C
1.000g/cm³
Liquid
7
Colorless
100°C
15, 10237
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02
Poly CUBE
Filtered through a 0.2-micron filter.
4 L

Chemical Identifiers

7732-18-5
18.015
dihydrogen oxide, dihydrogen monoxide
CHEBI:15377
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
Videos
Safety and Handling

Safety and Handling

Low hazard for usual industrial or commercial handling.

Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult

SDS Documents

Documents

Promotions

Promotions