missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, CHROMASOLV™, Honeywell Riedel-de Haën™

CHROMASOLV™, for pesticide residue analysis

Supplier:  Honeywell Chemicals 344786X1L

 View more versions of this product

Catalog No. 6002514


Explore available promotions

Description

Description

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
Specifications

Specifications

Water
H2O
NONH for all modes of transport
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
O
18.015
CHEBI:15377
CHROMASOLV™
6 x 1 L
7732-18-5
MFCD00011332
2050024
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02g/mol
1.000g/mL at 3.98°C

Chemical Identifiers

7732-18-5
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
H2O
MFCD00011332
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
CHEBI:15377
O
Videos
Safety and Handling

Safety and Handling

RTECSNumber : ZC0110000

SDS
Documents

Documents

Promotions

Promotions