missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Water, Distilled, Reagent Grade, Ricca Chemical
Supplier: Ricca Chemical Company 91801
Specifications
Water | |
Colorless | |
max. 1.5 μS/cm | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
Amber Glass Bottle | |
4 L |
Distilled | |
7732-18-5 | |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
Reagent | |
Non-sterile |
Chemical Identifiers
7732-18-5 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
CHEBI:15377 |
H2O | |
962 | |
O |
Safety and Handling
ShelfLife : 18 months