missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, DIUF, Fisher Chemical™

$252.83 - $601.50

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
W2-4
SDS View Documents
Thermo Fisher Scientific
W24
4 L Poly Bottle
Each for $252.83
Case of 4 Each for $1,011.32
 
W2-20
SDS View Documents
Fisher Chemical
FLW220
20 L PolyPac™
Each for $601.50
 
Description

Description

Water, DIUF

Specifications

Chemical Identifiers

7732-18-5
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
H2O
MFCD00011332
dihydrogen oxide, dihydrogen monoxide
CHEBI:15377
O

Specifications

7
7732-18-5
100°C
MFCD00011332
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02
17.5mmHg at 20°C
4 L
Water, D.I.U.F.
Colorless
0°C
H2O
dihydrogen oxide, dihydrogen monoxide
O
18.015
CHEBI:15377
Poly Bottle
1.000g/cm³
Liquid
Videos
Safety and Handling

Safety and Handling

SDS
missing translation for 'documents'

missing translation for 'documents'