Learn More
Water, (DNASE, RNASE free), Fisher BioReagents™
Water, H2O, CAS Number-7732-18-5, dihydrogen monoxide, dihydrogen oxide, 100mL, Vapor Pressure: 17.5mmHg at 20 deg.C, 100 deg.C, CHEBI:15377, Gray, Colorless, 1.000g/cm3, DNase free, Filtered through a 0.2-micron filter., 18.02, Low hazard for usual industrial or commercial handling.
Supplier: Fisher BioReagents BP2484100
Description
Water, H2O, CAS Number-7732-18-5, dihydrogen monoxide, dihydrogen oxide, 100mL, Vapor Pressure: 17.5mmHg at 20 deg.C, 100 deg.C, CHEBI:15377, Gray, Colorless, 1.000g/cm3, DNase free, Filtered through a 0.2-micron filter., 18.02, Low hazard for usual industrial or commercial handling.
Diethylpyrocarbonate (DEPC)-treated Water is guaranteed nuclease-free (tested for both DNase and RNase) and is suitable for all RNA work.Specifications
Water | |
DNASE, RNASE free | |
Colorless | |
100°C | |
H2O | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
Molecular Biology | |
17.5mmHg at 20°C | |
DEPC-Treated | |
100 mL |
7 | |
7732-18-5 | |
0°C | |
DNase free | |
15, 10237 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02 | |
Poly Bottle | |
Filtered through a 0.2-micron filter. | |
1.000g/cm³ | |
Liquid |
Chemical Identifiers
7732-18-5 | |
18.015 | |
dihydrogen oxide, dihydrogen monoxide | |
CHEBI:15377 | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
Safety and Handling
Low hazard for usual industrial or commercial handling.
Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult