missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Water, Extra Pure, Deionized, Thermo Scientific Chemicals
$82.16 - $344.13
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.015 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | dihydrogen oxide, dihydrogen monoxide |
PubChem CID | 962 |
ChEBI | CHEBI:15377 |
IUPAC Name | oxidane |
SMILES | O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC345470050
|
Thermo Scientific Chemicals
345470050 |
5 L | Plastic Drum |
Each for $82.16
|
|
||||
AC345470250
|
Thermo Scientific Chemicals
345470250 |
25 L | Plastic Drum |
Each for $344.13
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
7732-18-5 | |
18.015 | |
dihydrogen oxide, dihydrogen monoxide | |
CHEBI:15377 | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
Specifications
7732-18-5 | |
100.0°C | |
H2O | |
16,383 | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
Extra Pure | |
25ppm max. | |
1.0000g/mL | |
1.3334 | |
Liquid |
Colorless | |
5.0μS/cm Max. | |
H2O | |
15, 10237 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02 | |
Plastic Drum | |
0.89 mPa.s (25°C) | |
5 L | |
1 | |
Water |
Safety and Handling
RUO – Research Use Only