missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, For HPLC and Spectrophotometry, Honeywell

Supplier:  Honeywell Chemicals AH3654

Catalog No. 6003013


Explore available promotions

Description

Description

Specifications

Specifications

0.01 AU max. at 200 nm, 0.005 AU max. at 250 nm, 0.01 AU max. at 190 nm
For HPLC and Spectrophotometry
100°C
H2O
2050024
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18
Glass Bottle
4 x 4 L
Water
7732-18-5
100.0000%
MFCD00011332
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
O
18.015
CHEBI:15377
HPLC
0.9971 g/cm3

Chemical Identifiers

7732-18-5
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
H2O
MFCD00011332
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
CHEBI:15377
O
Videos
Safety and Handling

Safety and Handling

ShelfLife : 547 days from date of manufacture

EINECSNumber : 231-791-2

SDS
Documents

Documents

Promotions

Promotions