missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Water, for HPLC gradient grade, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 268300025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLCSpecifications
Water | |
7732-18-5 | |
100.0°C | |
H2O | |
16,383 | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
HPLC Gradient | |
17.5mmHg at 20°C | |
1.0000g/mL | |
1.3334 |
For HPLC gradient grade | |
Colorless | |
H2O | |
MFCD00011332 | |
15, 10237 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02 | |
0.0003% max. | |
0.89 mPa.s (25°C) | |
2.5 L | |
1 |
Chemical Identifiers
7732-18-5 | |
18.015 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
H2O | |
MFCD00011332 | |
dihydrogen oxide, dihydrogen monoxide | |
CHEBI:15377 | |
O |
Safety and Handling
EINECSNumber : 231-791-2
RTECSNumber : ZC0110000
TSCA : TSCA
RUO – Research Use Only