missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Water, HPLC, J.T. Baker™
High purity solvents for use in analytical and preparative separations
Supplier: Avantor J.T.Baker 421803
Description
- For use in liquid chromatography (HPLC & UHPLC) & spectrophotometry; For laboratory,research,or manufacturing use
Specifications
Water | |
7732-18-5 | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
Glass container | |
1L = 1.00kg |
Colorless | |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18 | |
4 L | |
Liquid |
Chemical Identifiers
7732-18-5 | |
18.015 | |
dihydrogen oxide, dihydrogen monoxide | |
CHEBI:15377 | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |