Water for Injection, EP/USP, Sterile Grade, Cytiva Hyclone™
Water for Injection, EP/USP, is a terminally distilled, sterile, non-pyrogenic preparation which contains no bacteriostat, antimicrobial agent or added buffer.
Supplier:  Cytiva Z0104
 View more versions of this product
Catalog No. Z0104
Explore available promotions
Description
Description
  • Complies with the USP and EP monographs for water for injection packaged in bulk for commercial use
  • Manufactured under cGMP and ISO 13485 guidelines in a FDA registered facility
  • Excellent choice for upstream and downstream media/buffer preparation in biopharmaceutical manufacturing applications
  • Widely used in medical device manufacturing/processing applications where a high quality, sterile water is required
Specifications
Specifications
Clear, Colorless
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
EP/USP
Sterile
4 x 4 L
7732-18-5
<0.25 EU/mL
O
18.02
HDPE Jug
Yes
Chemical Identifiers
7732-18-5
18.02
water
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
O
Videos
Safety and Handling
Safety and Handling
ShelfLife : 36 Months
Recommended Storage : 2-30°C