missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, CHROMASOLV™ LC-MS, Honeywell Riedel-de Haën™

CHROMASOLV™ LC-MS

Supplier:  Honeywell-Chromasolv 392531L

 View more versions of this product

Catalog No. 60046744


Explore available promotions

Description

Description

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities
Specifications

Specifications

max. 0.022 at 210 nm, max. 0.004 at 230 nm
CHROMASOLV™ LC-MS
100°C
H2O
MFCD00011332
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
O
18.015
CHEBI:15377
LC-MS
1.000 g/mL at 3.98 °C(lit.)
Water
7732-18-5
100.0000%
H2O
2050024
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18
Glass Bottle
1 L

Chemical Identifiers

7732-18-5
MFCD00011332
962
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
CHEBI:15377
Videos
Safety and Handling

Safety and Handling

ShelfLife : 547 days from date of manufacture

EINECSNumber : 231-791-2

SDS
Documents

Documents

Promotions

Promotions