missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, LC-MS Grade, Thermo Scientific Chemicals

Used as mobile phase solvent in LC-MS analysis

Supplier:  Thermo Scientific Chemicals 047146Y0

 View more versions of this product

Catalog No. AA47146Y0


Explore available promotions

Description

Description

It is used as mobile phase solvent in LC-MS analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Water
7732-18-5
0°C
H2O
2050024
Soluble in water; Insoluble in polar liquids.
O
18.015
CHEBI:15377
LC-MS
1 g/cm3
1.34
7 (at 20°C)
Colorless
100°C
MFCD00011332
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02
Odorless
6 x 1 L
Liquid

Chemical Identifiers

7732-18-5
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
H2O
MFCD00011332
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
CHEBI:15377
O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-791-2

RTECSNumber : ZC0110000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only