missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, Molecular Biology Grade, Fisher BioReagents

For many fundamental molecular biology procedures

Supplier:  Fisher BioReagents BP281910

 View more versions of this product

Catalog No. BP281910


Explore available promotions

Description

Description

Ultrapure, deionized, molecular biology grade water is ideal for many fundamental procedures such as PCR, electrophoresis, DNA sequencing, and buffers for enzymatic analyses.

Specifications

Specifications

Water
0.03μm filtered to ensure high purity
Colorless
100°C
<2μS/cm
H2O
dihydrogen oxide, dihydrogen monoxide
O
18.015
CHEBI:15377
Molecular Biology
17.5mmHg at 20°C
Deionized
10 L
7
7732-18-5
0°C
DNase-, RNase- and Protease-Free
DNase free
15, 10237
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02
Poly CUBE
Filtered through a 0.3μ filter
1.000g/cm³
Liquid

Chemical Identifiers

7732-18-5
18.015
dihydrogen oxide, dihydrogen monoxide
CHEBI:15377
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
Videos
Safety and Handling

Safety and Handling

Low hazard for usual industrial or commercial handling.

Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention. . . . .

NFPA

Health:0
Flammability:0
Instability:0

SDS Documents

Documents

Promotions

Promotions