missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

Water, Molecular Biology Grade, Fisher BioReagents
For many fundamental molecular biology procedures
Supplier: Fisher BioReagents BP2819100
Description
Ultrapure, deionized, molecular biology grade water is ideal for many fundamental procedures such as PCR, electrophoresis, DNA sequencing, and buffers for enzymatic analyses.
Specifications
Water | |
0.03μm filtered to ensure high purity | |
Colorless | |
100°C | |
<2μS/cm | |
H2O | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
Molecular Biology | |
17.5mmHg at 20°C | |
Deionized | |
100 mL |
7 | |
7732-18-5 | |
0°C | |
DNase-, RNase- and Protease-Free | |
DNase free | |
15, 10237 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02 | |
Poly Bottle | |
Filtered through a 0.3μ filter | |
1.000g/cm³ | |
Liquid |
Chemical Identifiers
7732-18-5 | |
18.015 | |
dihydrogen oxide, dihydrogen monoxide | |
CHEBI:15377 | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
Safety and Handling

Low hazard for usual industrial or commercial handling.
Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention. . . . .
NFPA
Health:0
Flammability:0
Instability:0