missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Water, Optima™ for HPLC, Fisher Chemical™
Supplier: Thermo Fisher Scientific W74
Description
Improved filtration! High purity mobile phase for HPLC
- Screened UV-absorbing contaminants to reduce interferences for smoother baselines
- Tested for HPLC gradient suitability
- Mobile phase for analytical and preparative separations
- Now filters through 0.2µm filter to remove particulates.
Specifications
0.005 AU max. at 254nm, 0.005 AU max. at 300nm, 0.005 AU max. at 400nm, 0.01 AU max. at 200nm | |
7 | |
Colorless | |
100°C | |
H2O | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
OPTIMA™ | |
1 ppm max. | |
1000 g/cm³ | |
4 L |
Water | |
7732-18-5 | |
0°C | |
High purity mobile phase for HPLC | |
MFCD00011332 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02 | |
Amber Soda Lime Glass | |
17.5mmHg at 20°C | |
Certificate of Lot Analysis included with each bottle; UV and other chromatograms available on request. | |
Liquid |
Chemical Identifiers
7732-18-5 | |
18.015 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
H2O | |
MFCD00011332 | |
dihydrogen oxide, dihydrogen monoxide | |
CHEBI:15377 | |
O |
Safety and Handling
Low hazard for usual industrial or commercial handling.
Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention. . . . .
NFPA
Health:0
Flammability:0
Instability:0