missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, Optima™ for HPLC, Fisher Chemical™

Supplier:  Thermo Fisher Scientific W7SK4

 View more versions of this product

Catalog No. W7SK-4



Explore available promotions

Description

Description

Improved filtration! High purity mobile phase for HPLC

  • Screened UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested for HPLC gradient suitability
  • Mobile phase for analytical and preparative separations
  • Now filters through 0.2µm filter to remove particulates.
Specifications

Specifications

0.005 AU max. at 254nm, 0.005 AU max. at 300nm, 0.005 AU max. at 400nm, 0.01 AU max. at 200nm
7
Colorless
100°C
H2O
dihydrogen oxide, dihydrogen monoxide
O
18.015
CHEBI:15377
OPTIMA™
1 ppm max.
1000 g/cm³
4 L
Water
7732-18-5
0°C
High purity mobile phase for HPLC
MFCD00011332
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02
Safe-Cote™ Amber Glass
17.5mmHg at 20°C
Certificate of Lot Analysis included with each bottle; UV and other chromatograms available on request.
Liquid

Chemical Identifiers

7732-18-5
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
H2O
MFCD00011332
dihydrogen oxide, dihydrogen monoxide
CHEBI:15377
O
Videos
Safety and Handling

Safety and Handling

Low hazard for usual industrial or commercial handling.

Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention. . . . .

NFPA

Health:0
Flammability:0
Instability:0

SDS Documents

Documents

Promotions

Promotions