missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, Reagent (Deionized water), ACS, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 036645K7

 View more versions of this product

Catalog No. AA36645K7


Explore available promotions

Description

Description

Used in conductometric titration, Synthesis of nanomaterials. Mostly it is used as solvent in industries and in all research fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Water
Colorless
100°C
MFCD00011332
Miscible in methanol,acetone,Ethanol.
O
18.015
CHEBI:15377
ACS
1
1.333
7732-18-5
0°C
H2O
dihydrogen oxide, dihydrogen monoxide
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02
Odorless
4 L
Liquid

Chemical Identifiers

7732-18-5
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
H2O
MFCD00011332
dihydrogen oxide, dihydrogen monoxide
CHEBI:15377
O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-791-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only