Thermo Scientific™ Water-Soluble Siliconizing Fluid
To siliconize glass surfaces use Thermo Scientific™ Water-Soluble Siliconizing Fluid.
Supplier:  Thermo Scientific™ TS42799
Catalog No. PI42799
Explore available promotions
Description
Description
To siliconize glass surfaces use Thermo Scientific™ Water-Soluble Siliconizing Fluid. This easy-to-use silane monomer solution is supplied as a 20% solid solution in a mixture of diacetone alcohol and tertiary butyl alcohol. The primary silane component is an octadecyltrialkoxysilane, which is hydrolyzed to a silanol when mixed with water. It provides greater resistance to base hydrolysis than other surface treatments.
  • Easy-to-use silane monomer solution that is supplied as a 20% solid solution in a mixture of diacetone alcohol and tertiary butyl alcohol
  • Greater resistance to base hydrolysis than other surface treatments
Specifications
Specifications
Siliconizing Glass Surfaces
4-Hydroxy-4-Methylpentan-2-One, 2-Methyl-2-Propanol, Silane, Trimethoxyoctadecyl
120 mL
Water-Soluble Siliconizing Fluid
Flammable, Irritant
25 - 45, 25 - 45, 10 - 20%
15°C to 35°C
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.