missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, SupraSolv™, EMD Milipore™
Supplier: Technidata 1005772500
Description
Recommended Storage
Storage temperature: no restrictions.
Specifications
| Colorless | |
| 100°C (1013Â hPa) | |
| dihydrogen oxide, dihydrogen monoxide | |
| O | |
| 18.015 | |
| CHEBI:15377 | |
| Glass Bottle | |
| 1.00Â g/3 (20Â °C) | |
| Liquid |
| 7732-18-5 | |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| oxidane | |
| 962 | |
| 18.02 | |
| 23Â hPa (20Â °C) | |
| 2.5 L |
Chemical Identifiers
| 7732-18-5 | |
| 18.015 | |
| dihydrogen oxide, dihydrogen monoxide | |
| CHEBI:15377 | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| 962 | |
| oxidane |
Safety and Handling
RTECSNumber : ZC0110000