missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, ULTREX™ II Ultrapure Reagent, J.T. Baker™

Reagents for critical elemental analysis with less than 10 parts-per-trillion (ppt) levels of up to 65 elements

Supplier:  Avantor 690602

Catalog No. 02004559

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

    For use in ICP (Inducutively Coupled Plasma Spectrometry), Trace Metal Analysis
Specifications

Specifications

Water
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18
Narrow Mouth Poly Bottle
1L = 1.00kg
7732-18-5
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
O
18.015
CHEBI:15377
ULTREX™ II Ultrapure Reagent
1 L

Chemical Identifiers

7732-18-5
18.015
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
CHEBI:15377
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
Videos
SDS
Documents

Documents

Promotions

Promotions