missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Water, USP/EP Purified, Ricca Chemical

Supplier:  Ricca Chemical Company 91901G

 View more versions of this product

Catalog No. 9190-1G


Explore available promotions

Description

Description

Water, USP/EP Purified

Specifications

Specifications

Water
Clear, Colorless
98.0
max. 1 μS/cm
max. 0.25 EU/mL at time of manufacture
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
EP Purified/USP
max. 0.001 %
4 L
5.0 to 7.0
7732-18-5
100.0
H2O
dihydrogen oxide, dihydrogen monoxide
O
18.015
CHEBI:15377
Amber Glass Bottle
Non-sterile

Chemical Identifiers

7732-18-5
XLYOFNOQVPJJNP-UHFFFAOYSA-N
CHEBI:15377
H2O
962
O
Videos
Safety and Handling

Safety and Handling

ShelfLife : 12 months

SDS Documents

Documents

Accessories
Promotions

Promotions