Wearwell™ Diamond-Plate SponeCote Work Mat with Gritshield Surface
Combines productivity improvement with slip prevention in oily overspray applications
Supplier:  Wearwell™ 416.916X2X3BK
 View more versions of this product
Catalog No. 19-130-3546
Explore available promotions
Description
Description
  • Nitrile enhanced Nitricell sponge base absorbs repeated compression
  • Available in Diamond-Plate with GRITSHIELD UltraSoft and GRITSHIELD surface
  • Machine cut and beveled edges prevent tripping
  • Maximized standing area
  • Overall Thickness: 15/16 in. (24mm)
Specifications
Specifications
2 x 3 ft.