Wearwell™ ESD Wrist Strap
Use wrist strap to ground workers to their ESD tabletop mats
Supplier: Wearwell™ 7936WSTRAP
Description
- Comfort strap with copper sulfide fibers
- Use with work surface mat
- Attached to a 6 ft. L coil cord w/snap on one end and banana jack on other
- Width: 2.54cm (1 in.) and w/1.8m (6 ft.) coil cord
Specifications
Coil Cord w/Wrist Strap | |
6 ft. | |
Black |